Learn More
Kartell™ Plastilab™ Conical Stoppers
Features tapered conical-shaped bottom to ensure sealing action.
Brand: Kartell™ 0124200
This item is not returnable.
View return policy
Description
- Made from Polyethylene for tight and secure seal
- Raised embossed easy-to-read sizes
- Color-coded
Specifications
DIN 12242 | |
NS 12/21 | |
10.4 mm | |
Tapered Stopper |
Conical bottom | |
Green | |
Polyethylene | |
Pentagonal Grip |