Learn More
Ibidi μ-Plate 96 Well 3D ibiTreat
μ-Plate 96 Well 3D ibiTreat is a black 96 well plate for 3D cell culture.
Brand:  Ibidi 89646
Encompass_Preferred
Product Code. 15193523
  • 618.31 € / Pack of 15
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • A black 96 well plate for high-throughput 3D cell culture and tube formation assays
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Specifications
Specifications
Black
Well Plate
96
15
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ibidi μ-Plate 96 Well 3D ibiTreat