Learn More
Thermo Scientific™ Oxoid™ Plate Count agar Cm325 Oxoid
Brand: Thermo Scientific™ Oxoid™ CM0325T
1873.10 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Straw powder | |
Powder | |
5 kg |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6251253 | Certificate of Analysis | 03/04/2025 | CM0325T |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Oxoid™ Plate Count agar Cm325 Oxoid >
Spot an opportunity for improvement?Share a Content Correction