Learn More
Memmert™ Plug-In Tube Extension for SN160, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand: Memmert™ SN160B29718
This item is not returnable.
View return policy
Description
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Specifications
60.3 mm | |
SN160 Sterilizer |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Plug-In Tube Extension for SN160, Straight > For Use With: SN160
Spot an opportunity for improvement?Share a Content Correction