Learn More
Memmert™ Plug-In Tube Extension for SN750, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand:  Memmert™ SN750B29719
Product Code. 17538715
  • 129.00 € / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Specifications
60.3 mm
SN750 Sterilizer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Plug-In Tube Extension for SN750, Straight > For Use With: SN750