missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Plug-In Tube Extension for UF450, Straight

Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.

Brand:  Memmert™ UF450B29719

Product Code. 17559325

  • 129.00 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications

Specifications

UF450 Oven
Plug-in Tube Extension
O.D. 60.3 mm, I.D. 57 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions