missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Plumbtesmo Qualitative Test Paper

Product Code. 10239981 Shop All Macherey-Nagel Products
Click to view available options
Color Indicator:
White/Yellow to Pink
Unit Size:
Pack of 40
This item is not returnable. View return policy

Product Code. 10239981

Brand: Macherey-Nagel™ 90602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Indicator White/Yellow to Pink
Format Sheet
For Use With (Application) For detection of Lead, lead ions
Quantity 40 Pack
Test Time 2 min.
Type Qualitative Test Paper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.