Learn More
Podocarpic acid, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Podocarpic acid is a natural product, which has the best all-round positive effect and acts as a novel TRPA1 activator.
Specifications
Specifications
| Chemical Name or Material | Podocarpic acid |
| CAS | 5947-49-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H22O3 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (364.50 mM) |
| SMILES | O=C([C@@]1(C)CCC[C@]2(C)C3=C(CC[C@@]12[H])C=CC(O)=C3)O |
| Molecular Weight (g/mol) | 274.35 |
| Formula Weight | 274.35 |
| Show More |