Learn More
Cole-Parmer™ PolarSafe™ Cryogenic Storage Dewars with Racks
Cole-Parmer™ PolarSafe™ Cryogenic Storage Dewars with Racks safely store samples for up to 200 days without liquid nitrogen refill
Brand:  Cole-Parmer™ 4426055
 View more versions of this product
Product Code. 16619551
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Liquid nitrogen dewars available in sizes from 2 L to 115 L
  • Ultra low evaporation loss enhances cryogen protection
  • Durable aluminum construction
  • Dual handles aid mobility
  • 5 year vacuum warranty
Specifications
Specifications
Static Holding Time: 80 Days
30 in.
8.5 in.
21.59 cm
Aluminum
115 L
76.2 cm
30 in.
76.2 cm