Learn More
Koflo Unmounted In-Line Static Mixers
Product Code. 11774109 Shop All Cole-Parmer Products
Click to view available options
Length (Metric):
121 mm
127 mm
152 mm
241 mm
254 mm
Unit Size:
Pack of 10
Product Code. 11774109
Brand: Koflo 0466718
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Unmounted In-Line Static Mixers can be used with any standard tubing having the same tubing ID as the element OD given in the table.
  • They work well with flexible tubing.
Specifications
Diameter (English) 0.5 in.
Diameter (Metric) 13 mm
Length (English) 10 in.
Length (Metric) 254 mm
Type Unmounted In-line Mixer