Learn More
Macherey-Nagel™ Polyester Sheets Polygram SIL G, Alox N, CEL 300/UV254 TLC Micro-Set M
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
4 x 8 cm
Unit Size:
Pack of 200
Description
Different TLC polyester sheets POLYGRAM in format 4x8 cm
Specifications
Specifications
| Material | Polyester Sheets Polygram SIL G, Alox N, CEL 300 |
| Quantity | 200 Pk. |
| Type | TLC Micro-Set M |
| Fluorescent Indicator | UV254 |
| Includes | 2 developing chambers; 2 x 50 glass capillaries 1 μL; 1 rubber cap for glass capillaries; 2 spotting guides; 1 measuring cylinder 10 mL; 1 beaker 25 mL; 1 glass laboratory sprayer with rubber bulb; 1 plastic syringe 1 mL; 20 Sheets filter paper MN 713, 15 x 21 cm; 50 polyester sheets Polygram SIL G/UV254, Alox N/UV254, CEL 300 |
| Dimensions (L x W) | 4 x 8 cm |