missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 6000, Carl Roth™

Product Code. 10181343
Encompass
Click to view available options
Quantity:
1 kg
Packaging:
Plastic
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 107-21-1
pH 5.0 to 7.0
Packaging Plastic
Molecular Formula C2H6O2
Quantity 1 kg
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, polyethylene glycol, macrogol
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Molecular Weight (g/mol) 62.07
PubChem CID 174
ChEBI CHEBI:30742
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'