Learn More
Hirschmann™ Polypropylene Thread Adapter
Polypropylene (PP) Threaded adapter for adaptation on the most common bottles.
10.35 € - 17.00 €
Product Code | Brand | Thread Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | ||||||
10380201
|
Hirschmann™
9313505 |
A 32 to A 38 |
15.20 €
Each |
In Stock
Log in to see stock. |
||||||
10009740
|
Hirschmann™
9313527 |
A 45 to S 40 |
16.10 €
Each |
In Stock
Log in to see stock. |
||||||
10319820
|
Hirschmann™
9313506 |
A 32 to A 45 |
17.00 €
Each |
In Stock
Log in to see stock. |
||||||
10195260
|
Hirschmann™
9313502 |
25 mm |
10.35 €
Each |
In Stock
Log in to see stock. |
||||||
10482101
|
Hirschmann™
9313503 |
28 mm |
10.35 €
Each |
In Stock
Log in to see stock. |
||||||
10624542
|
Hirschmann™
9313521 |
32 mm |
16.10 €
Each |
In Stock
Log in to see stock. |
||||||
10408372
|
Hirschmann™
9313525 |
38 mm |
16.10 €
Each |
In Stock
Log in to see stock. |
||||||
10278130
|
Hirschmann™
9313507 |
40 mm |
16.10 €
Each |
In Stock
Log in to see stock. |
||||||
10495243
|
Hirschmann™
9313524 |
38/430 mm |
16.40 €
Each |
In Stock
Log in to see stock. |
||||||
Description
- 9313505/9313506: For use with ceramus™ -classic.
- 9313527: For use with ceramus™, solarus™ and opus™.
Specifications
Adapter | |
Polypropylene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hirschmann™ Polypropylene Thread Adapter