Learn More
BRAND™ Polypropylene Funnel with Internal Fluting
Product Code. 10633371 Shop All Brand Products
Click to view available options
Diameter (Metric) Stem:
10 mm
Unit Size:
Pack of 20
Product Code. 10633371
Brand: BRAND™ 148225
Only null left
Add to basket
Add to basket
Polypropylene Funnel with internal fluting and a short, wide stem.
  • Includes a short and wide stem
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 80 mm
Diameter (Metric) Stem 10 mm
Material Polypropylene
Includes Internal Fluting
Length (Metric) Overall 69 mm
Product Type Funnel