Learn More
Fisherbrand™ Polypropylene Graduated Microcentrifuge Tubes
Use these tubes for a variety of applications. Fisherbrand™ Polypropylene Graduated Microcentrifuge Tubes are ideal for use with PCR systems and centrifugation.
Brand: Fisherbrand™ FB74061
This item is not returnable.
View return policy
Description
- Polypropylene graduated tubes
- Autoclavable
- Available in a variety of colors for easy identification
- Available with pick-up tab
Specifications
Autoclavable | |
Natural | |
For PCR, Centrifugation | |
Yes |
1.5 mL | |
Polypropylene | |
500/Pk. | |
Snap Cap |