missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Polypropylene Reagent Tube Racks
Brand: VITLAB™ 80562
Additional Details : Weight : 0.01000kg
Specifications
Reagent Tube Rack | |
Polypropylene | |
5 Pack | |
21 mm |
Tubes (21 mm) | |
24 | |
-20°C to +90°C |