Learn More
Bohlender™ Polypropylene Sampling Pump
Click to view available options
Flow Rate:
60 mL
Unit Size:
Each
Specifications
Specifications
| Flow Rate | 60 mL |
| For Use With | 100 to 2000 mL Bottle |
| Material | Polypropylene, PTFE |