Learn More
Macherey-Nagel™ Polypropylene Screw Caps with Hole
Product Code. 12725232 Shop All Macherey-Nagel Products
Click to view available options
Color:
Green
Unit Size:
Each
Product Code. 12725232
Brand: Macherey-Nagel™ 702082
Please to purchase this item. Need a web account? Register with us today!
Polypropylene closures with red rubber/FEP septum for routine analysis and a fair price-performance-ratio. Easy and safe needle penetration. Compatible with Agilent, Knauer autosamplers and GC/HPLC applications.
  • Green color
  • Red rubber/colorless FEP septum
  • For GC and HPLC applications
  • Pre-assembled
Specifications
Color (Seal) Red Rubber/Colorless FEP
Description PP Screw Cap with Hole and Pre-Fitted Septa
Color Green
Material Polypropylene
Septum Red Rubber/Colorless FEP
For Use With (Application) GC and HPLC
Temperature Range (Metric) Temp. Resistance -40°C up to 110°C/Analytical Purity - Medium/Medium Hard to Penetrate/Medium Resealability
Type Screw Cap
For Use With (Equipment) Agilent and Knauer Autosamplers