Learn More
Lenz Laborglasinstrumente™ Plastic Straight Hose Connector
For connecting screw caps with bore. Lenz Laborglasinstrumente™ Plastic Straight Hose Connectors are made from polypropylene.
Brand: Lenz Laborglasinstrumente™ 01350000
This item is not returnable.
View return policy
Description
- Autoclavable
- Available in two different thread sizes
- Silicone sealing
Specifications
Autoclavable | |
For screw caps with bore | |
GL14 | |
Straight Hose Connection |
Polypropylene | |
10 Pack | |
Straight |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasinstrumente™ Plastic Straight Hose Connector > Thread Size: GL14
Spot an opportunity for improvement?Share a Content Correction