Learn More
Greiner Bio-One Polystyrene Conical Bottom Analyzer Cups
Greiner Bio-One Cell 1.5mL Polystyrene Conical Bottom Analyzer Cups suitable for use with analytical systems, such as Technicon, Gemsaec, Centrifichem, Hitachi, TOA and Coulter analyzers.
Brand: Greiner Bio-One 621171
This item is not returnable.
View return policy
Description
- Material: Polystyrene
- Conical bottom
- With support skirt
- For single use only
Specifications
Technicon Analyzer | |
Non-sterile | |
Polystyrene | |
4 mL |
Conical | |
Clear | |
Analyzer Cup |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One Polystyrene Conical Bottom Analyzer Cups > Suitable for Technicon analyzer; Volume: 4mL
Spot an opportunity for improvement?Share a Content Correction