missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems Porcine Luminex™ Discovery Assay

Product Code. 18732024
Click to view available options
Quantity:
1 Plex
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18732024

Brand: R&D Systems LXSAPM09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 9 Plex
Species Porcine
Binding Target Multiplex Cytokine Assay Kits
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.