missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Portable pH/mV Meter

Features automatic calibration at one or two points and ATC when using the temperature probe. Hanna Instruments™ Portable pH/mV Meter is a compact, waterproof meter supplied complete with a protective carrying case, pH electrode, temperature probe, calibration buffer and cleaning solution.

Brand:  Hanna Instruments™ HI8424N

Product Code. 10031522

  • 592.00 € / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This economical portable pH/ mV meter uses a BNC connector for the pH electrode input, allowing for a wide selection of pH and ORP electrodes to be used. A separate temperature probe is supplied to provide temperature compensated pH measurement. The HI8424 is very basic in operation, with automatic calibration to any of the three programmed buffer values. The simplicity and the economical price has made it one of the most popular meters available in the market.

  • Waterproof: Compact, heavy duty casing protects against water that would damage the electronics
  • Automatic Calibration: Calibration to one or two points to programmed buffer values (pH 4.01, 7.01, and 10.01)
  • Automatic Temperature Compensation: Separate temperature probe is supplied for temperature compensated measurements
  • High Resolution/Accuracy mV Range: Automatically switches resolution from 1 mV to 0.1 mV when readings are between -699.9 to +699.9 mV
  • Automatic Shutoff: Turns the meter off after 20 minutes of disuse to conserve battery life
  • Low Battery Indicator: Displays when the battery life is almost exhausted (5% remaining) for timely replacement
Specifications

Specifications

HI1230B pH electrode, HI7662 temperature probe, HI70004 pH 4.01 buffer solution sachet, HI70007 pH 7.01 buffer solution sachet, HI700601 electrode cleaning solution sachets (2), battery, protective case and instructions
±0.02, ±1
Automatic
0.01
0.1 mV, 1 mV
-20.0°C to +120.0°C
0.1°C/F
180 g
2 Years (6 Months Probe)
HI1230B pH probe with BNC connector and 1m cable, HI7662 temperature probe with 1m cable
Automatic from -20.0 to 120.0°C with temperature probe attached, or manual
0°C to 50°C
150 hr. (auto-off: 20 min.)
±0.4°C/±0.8°F
±0.01
-2.00 to 16.00
±699.9 to ±1999
1 or 2
-4.0°F to +248.0°F
164 x 76 x 45 mm (6.5 x 3.0 x 1.8 in.)
6.3 oz.
pH electrode: PEI body, temperature probe: stainless steel
Max 100%
32°F to 122°F
9V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.