missing translation for 'onlineSavingsMsg'
Learn More
Potassium 3-nitrophenyltrifluoroborate, 95%, Thermo Scientific™ Product Code.: 15454297

Potassium 3-nitrophenyltrifluoroborate, 95%, Thermo Scientific™

Product Code. 15454297
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15454297

Brand: Thermo Scientific Alfa Aesar H31684.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 192863-40-4
Molecular Formula C6H4BF3KNO2
MDL Number MFCD03701627
Quantity 1 g
Synonym potassium 3-nitrophenyl trifluoroborate, potassium 3-nitrophenyltrifluoroborate, potassium trifluoro 3-nitrophenyl boranuide, potassium trifluoro 3-nitrophenyl borate, potassium trifluoro-3-nitrophenyl boranuide, pubchem11373, potassium ion trifluoro 3-nitrophenyl boranuide, 3-nitrophenyltrifluoroborate potassium salt, potassium tris fluoranyl-3-nitrophenyl boranuide
InChI Key ZKZMSCLFLMORNI-UHFFFAOYSA-N
SMILES [K+].[O-][N+](=O)C1=CC=CC(=C1)[B-](F)(F)F
IUPAC Name potassium;trifluoro-(3-nitrophenyl)boranuide
Molecular Weight (g/mol) 229.01
PubChem CID 2782884
Formula Weight 229.01
Percent Purity 95%
Chemical Name or Material Potassium 3-nitrophenyltrifluoroborate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10162539 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium 3-nitrophenyltrifluoroborate, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.