missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7073452 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
This item is not returnable. View return policy

Product Code. 10508070

Brand: Fisher Chemical P/7240/50

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number 11413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Melting Point 170°C
Color Colorless to White
pH 5.3 to 8.7
Physical Form Crystalline Solid
Quantity 250 g
Formula Weight 97.18g/mol
Packaging Plastic powder jar
Vapor Pressure <1 hPa at 20°C
compliance-icons
Product Identifier
  • Potassium thiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P233-Keep container tightly closed.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.