Learn More
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UF75
Potential-free contact for combination error message (e.g. supply failure, sensor fault, fuse).
Brand:  Memmert™ UF75H6
Product Code. 17519675
  • 298.00 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • For combination error message
Specifications
Specifications
UF75 Oven
Socket to NAMUR NE 28
2 A
Potential-free Contact
24 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UF75 > For Use With: UF75