Learn More
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UN160plus
Shop All Memmert Products
Click to view available options
Amperage:
2 A
Unit Size:
Each
Description
- Accessory
- For combination error message
Specifications
Specifications
| For Use With (Equipment) | UN160plus Oven |
| Type | Potential-free Contact |
| Includes | Socket to NAMUR NE 28 |
| Voltage | 24 V |
| Amperage | 2 A |