Learn More
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UN160plus
Product Code. 17549935 Shop All Memmert Products
Click to view available options
Amperage:
2 A
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 17549935
Brand: Memmert™ UN160plusH6
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Potential-free contact for combination error message (e.g. supply failure, sensor fault, fuse).
  • Accessory
  • For combination error message
Specifications
For Use With (Equipment) UN160plus Oven
Type Potential-free Contact
Includes Socket to NAMUR NE 28
Voltage 24 V
Amperage 2 A