Learn More
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UN260plus
Potential-free contact for combination error message (e.g. supply failure, sensor fault, fuse).
Brand: Memmert™ UN260plusH6
This item is not returnable.
View return policy
Description
- Accessory
- For combination error message
Specifications
UN260plus Oven | |
Socket to NAMUR NE 28 | |
2 A |
Potential-free Contact | |
24 V |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UN260plus > For Use With: UN260plus
Spot an opportunity for improvement?Share a Content Correction