Learn More
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Signal Generation for UN160plus
Potential-free contact for signal generation, controlled by program segment, for free-selectable functions to be activated.
Brand: Memmert™ UN160plusH72
Description
- Accessory
- For signal generation
- For free-selectable functions to be activated (e.g. activation of audible and visual signals, exhaust motors, fans, stirrers, etc.)
- Only for units with TwinDISPLAY
- Max. 2 contacts on 1-phase appliances
- Max. 4 contacts on 3-phase appliances
Specifications
UN160plus Oven | |
Socket to NAMUR NE 28 | |
2 A |
Potential-free Contact | |
24 V |