Learn More
Kern Sohn Precision Balance EG
The classic balance with robust tuning fork measuring system.
Encompass_Preferred
1132.20 € - 1326.00 €
Specifications
Type Precision Balance
Electrical Requirements 230 V, 50 Hz
View More Specs
Products 4
Product Code Brand Capacity Price Quantity & Availability  
Product Code Brand Capacity Price Quantity & Availability  
17341573
Encompass_Preferred
View Documents
Kern Sohn
EG 2200-2NM
2200 g
1132.20 €
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17361573
Encompass_Preferred
View Documents
Kern Sohn
EG 4200-2NM
4200 g
1244.40 €
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17351573
Encompass_Preferred
View Documents
Kern Sohn
EG 420-3NM
420 g
1183.20 €
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17371573
Encompass_Preferred
View Documents
Kern Sohn
EG 620-3NM
620 g
1326.00 €
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • Internal adjustment in the case of a change in temperature and time-controlled at defined intervals, guarantees high degree of accuracy and makes the balance independent of its location of use
  • Stable temperature behavior
  • Short stabilization time
  • Shock proof construction
  • High corner load performance
  • GLP/ISO record keeping of weight values
  • Totalizing of pieces when counting
  • Protective working cover included with delivery
Specifications
Specifications
Precision Balance
230 V, 50 Hz
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Kern Sohn Precision Balance EG