Learn More
Kern Sohn Precision Balance EG
The classic balance with robust tuning fork measuring system.
1132.20 € - 1326.00 €
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
17341573
![]() |
Kern Sohn
EG 2200-2NM |
2200 g |
1132.20 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17361573
![]() |
Kern Sohn
EG 4200-2NM |
4200 g |
1244.40 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17351573
![]() |
Kern Sohn
EG 420-3NM |
420 g |
1183.20 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17371573
![]() |
Kern Sohn
EG 620-3NM |
620 g |
1326.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Internal adjustment in the case of a change in temperature and time-controlled at defined intervals, guarantees high degree of accuracy and makes the balance independent of its location of use
- Stable temperature behavior
- Short stabilization time
- Shock proof construction
- High corner load performance
- GLP/ISO record keeping of weight values
- Totalizing of pieces when counting
- Protective working cover included with delivery
Specifications
Precision Balance | |
230 V, 50 Hz |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Kern Sohn Precision Balance EG