Learn More
Invitrogen™ PRIMER, 5'ALEXA 532 50 NMOL
Custom Primer
Brand:  Invitrogen™ 10660001
120.25 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 10413212
  • 185.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5' Alexa 532 Primer, 50 nmol
5' Alexa 532 Primer
5' Labeled
50 nmol