Learn More
Invitrogen™ PRIMER, 5'ALEXA 546 200 NMOL
Custom Primer
Brand: Invitrogen™ 10660010
This item is not returnable.
View return policy
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
5' Alexa 546 Primer, 200 nmol | |
5' Alexa 546 Primer | |
5' Labeled | |
200 nmol |