Learn More
Invitrogen™ PRIMER, 5'ALEXA 750 50 NMOL
Custom Primer
Brand:  Invitrogen™ 10660007
141.75 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10462252
  • 189.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
This item is not returnable. View return policy
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5' Alexa 750 Primer, 50 nmol
5' Alexa 750 Primer
5' Labeled
50 nmol