Learn More
Invitrogen™ PRIMER 5' HEX 50 NMOL
Custom Primer
Brand:  Invitrogen™ 11862018
81.00 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10580323
  • 108.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5' HEX Primer, 50 nmol
5' HEX Primer
5' Labeled
50 nmol