Learn More
Invitrogen™ Primer 5' OGC 488
Custom Primer
Brand: Invitrogen™ 11763026
This item is not returnable.
View return policy
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
5' OGC 488 Primer, 25 nmol | |
5' OGC 488 Primer | |
5' Labeled | |
25 nmol |