Learn More
Ansell™ VersaTouch™ 92-465 Series Blue Nitrile Splash Gloves
Provides outstanding resistance to animal fats compared with PVC or natural rubber. Ansell VersaTouchTM 92-465 Series Blue Nitrile Splash Gloves are manufactured from an innovative nitrile compound which offers excellent chemical and mechanical properties, comfort and dexterity.
Brand: Ansell™ 92-465_9.5-10
This item is not returnable.
View return policy
Description
- Contains no natural rubber
- Suitable for contact with all fatty foods
- Textured finish on fingertips for an improved grip
- Powder-free - no risk of contaminating food with corn startch used to powder gloves
- Ideal for all final stages of meat and fish processing, controlled environment rooms, dairy processing and catering
Specifications
EN 420:2003 + I1:2009 and EN 374:2003 | |
Nitrile | |
4.7 mil | |
9246510 |
Fully Textured | |
10 | |
0.12 mm | |
92-465 |