Learn More
Propargite, Honeywell Fluka™
Brand: Honeywell Fluka R1712-100MG
Additional Details : Weight : 0.10000kg ??product.attributeSet.Transport?? : UN number : 2810 Chem class : 6.1 Pack group : II
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Propargite | |
≤ 100% | |
2-(4-tert-Butylphenoxy)cyclohexyl 2-propynyl sulfite | |
1.12 g/cm3 (20°C) | |
Light Brown | |
71.4°C |
2312-35-8 | |
6934289 | |
350.47 g/mol | |
210°C (Decomposition) | |
100 mg |