Learn More
PROTAC ERα Degrader-2, MedChemExpress
MedChemExpress PROTAC ERα Degrader-2 comprises a IAP ligand binding group, a linker and an estrogen receptor α (ERα) binding group. PROTAC ERα Degrader-2 is an ERα degrader. Maximal ERα degradation at 30 μM concentration in human mammary tumor MCF7 cells. Degradation inducers based on cIAP1 are called specific and non-genetic IAP-dependent protein erasers (SNIPERs).
Brand: Medchem Express HY-111846
Description
PROTAC ERα Degrader-2 comprises a IAP ligand binding group, a linker and an estrogen receptor α (ERα) binding group. PROTAC ERα Degrader-2 is an ERα degrader. Maximal ERα degradation at 30 μM concentration in human mammary tumor MCF7 cells. Degradation inducers based on cIAP1 are called specific and non-genetic IAP-dependent protein erasers (SNIPERs).Specifications
PROTAC ERα Degrader-2 | |
White | |
C42H61N5O8 | |
DMSO : 200 mg/mL (261.79 mM; Need ultrasonic) | |
763.96 | |
98.02% | |
Solid |
1351169-29-3 | |
Research | |
5 mg | |
C[C@@]12[C@](CC/C2=N\OCC(NCCOCCOCCNC([C@H](CC(C)C)NC([C@@H](O)[C@H](N)CC3=CC=CC=C3)=O)=O)=O)([H])[C@@]4([H])[C@](C5=CC=C(O)C=C5CC4)([H])CC1 | |
763.96 | |
Research | |
Cancer-programmed cell death |