missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protein Precipitation Buffer MP
Additional protein precipitation buffer for use with NucleoSpin™ miRNA kits for the isolation of total RNA (small and large RNA) from diverse sample materials.
49.80 € - 152.00 €
Specifications
Color | Colorless |
---|---|
For Use With (Equipment) | NucleoSpin miRNA kit |
pH | 4.0 to 4.5 |
CAS | 127-09-3 |
Odor | Acetic |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15819097
|
Macherey-Nagel™
740407.20 |
20 mL (for 50 preps) |
49.80 €
20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15830908
|
Macherey-Nagel™
740407.100 |
100 mL (for 250 preps) |
152.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NucleoSpin™ miRNA Kit for small and large RNA isolation
- Excellent total RNA recovery and purity by chaotropic salt lysis
- Convenient extraction without phenol/chloroform
- Support protocol for fractionation of RNA by size in two separate fractions
- Small RNA (< 200 nt) and Large RNA (> 200 nt)
- Additional isolation of total protein fraction ready to use for SDS-PAGE and Western blot analysis
- NucleoSpin™ Filters for efficient sample homogenization
- rDNase for efficient on-column removal of genomic DNA
Specifications
Colorless | |
4.0 to 4.5 | |
Acetic | |
Soluble in Water | |
Protein Precipitation Buffer MP |
NucleoSpin miRNA kit | |
127-09-3 | |
Room Temperature (18°C to 25°C) | |
RNA Isolation |
Safety and Handling
missing translation for 'shelfLife' : At least 1 year
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Buffer MP (20 mL)
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : zinc chloride