Learn More
Thermo Scientific™ Proteome Discoverer™ Software
Product Code. 18897301 Shop All Thermo Scientific Products
Click to view available options
Includes:
XlinkX nodes
Unit Size:
Each
This item is not returnable. Vea la política de devoluciones
Código de producto. 18897301
missing translation for 'mfr': Thermo Scientific™ B51001481
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
Este artículo no se puede devolver. Vea la política de devoluciones
Simplify and enable a wide range of proteomics workflows, from identification to PTM analysis to isobaric mass tagging for quantitation with intelligent innovations.
Identify and quantify peptides and proteins in complex biological samples using Thermo Scientific™ Proteome Discoverer™ software. Proteome Discoverer software simplifies a wide range of proteomics workflows, from protein and peptide identification to PTM analysis to isobaric mass tagging and both SILAC and label-free quantitation. It supports multiple database search algorithms and multiple dissociation techniques. Go beyond searching to discovering with intelligent INFERYS™ Rescoring and the CHIMERYS™ intelligent search algorithm by MSAID to substantially increase the number of unique peptide identifications.
Proteome Discoverer software supports multiple dissociation techniques, quantitation technologies, and database search algorithms for comprehensive protein identification, characterization, and quantitation.
Comprehensive
  • Provides a choice of search algorithms (SEQUEST™ HT, Mascot, MS Amanda, Byonic™, ProsightPD™) and can combine output from multiple algorithms to maximize and cross-validate results
  • Find more with the intelligent INFERYS Rescoring and CHIMERYS algorithms
  • Supports multiple dissociation techniques (HCD, CID, ETD, and UVPD) for more complete coverage and confident identification of PTMs
  • Supports isobaric mass tagging (TMT, iTRAQ), isotopic labeling (SILAC), and label-free quantification
  • Validates peptide and protein IDs with false discovery rate (FDR)
Productive and Extensible
  • Automates data analysis and simplifies MSn data analysis through pre-built and customizable workflows
  • Includes study management for mapping of quantitation to biological information about the sample
  • Use results to refine data collecting, such as exporting inclusion and exclusion lists for import directly into instrument methods
  • Illustrates biological context by automatically annotating identified proteins with GO classifications, PTM sites, and literature references from public databases
  • Facilitates data comparison, exchange, and verification with support for data standards developed by the HUPO Proteomics Standards Initiative (PSI)
  • Supports third party nodes for data processing and enables processing of large datasets
  • Fast, robust algorithms empowered by artificial intelligence for peak selection and peptide quantification to extract more information
  • Obtain reliable statistics and validate quantification results visually
Especificaciones
Type Discoverer 3.1 XlinkX nodes
Software 3.1
For Use With (Equipment) Mass Spectrometers
Includes XlinkX nodes
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado