Learn More
PSI-7409 tetrasodium, MedChemExpress
Product Code. 16493486
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
Product Code. 16493486
Brand: Medchem Express HY15745A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress PSI-7409 tetrasodium is an active 5'-triphosphate metabolite of sofosbuvir (PSI-7977), inhibiting HCV NS5B polymerases, with IC50s of 1.6, 2.8, 0.7 and 2.6 μM for GT 1b_Con1, GT 2a_JFH1, GT 3a, and GT 4a NS5B polymerases, respectively.
PSI-7409 tetrasodium is an active 5'-triphosphate metabolite of sofosbuvir (PSI-7977), inhibiting HCV NS5B polymerases, with IC50s of 1.6, 2.8, 0.7 and 2.6 μM for GT 1b_Con1, GT 2a_JFH1, GT 3a, and GT 4a NS5B polymerases, respectively.
Specifications
Chemical Name or Material PSI-7409 tetrasodium
CAS 1621884-22-7
Color White
Purity Grade Notes Research
Molecular Formula C10H12FN2Na4O14P3
Quantity 25 mg
Solubility Information H2O : ≥ 125 mg/mL (212.55 mM)
SMILES O[C@@H]([C@@](C)(F)[C@H](N1C(NC(C=C1)=O)=O)O2)[C@H]2COP(OP(OP(O[Na])(O[Na])=O)(O[Na])=O)(O[Na])=O
Molecular Weight (g/mol) 588.09
Formula Weight 588.09
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus