Learn More
Quickfit™ PTFE Joint Clips with Conical Joints
Highly resistant to chemical attack
Brand: Quickfit GC45
87.92 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
Highly resistant to chemical attack
- Used for retaining glassware having Quickfit™ conical joints
- Manufactured from PTFE with encapsulated steel spring to provide firm grip
- Clips are numbered with joint size for easy identification
- Can be used at temperatures up to 250°C
Specifications
Joint Clip | |
Encapsulated Steel spring | |
45/40 |
PTFE | |
Conical Joints | |
250°C |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Quickfit™ PTFE Joint Clips with Conical Joints > Joint Size: 45/40
Spot an opportunity for improvement?Share a Content Correction