Learn More
Quickfit™ PTFE Joint Clips with Conical Joints
Highly resistant to chemical attack
Brand:  Quickfit GC24
34.25 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 12069062
  • 37.95 € / Pack of 5
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Highly resistant to chemical attack
  • Used for retaining glassware having Quickfit™ conical joints
  • Manufactured from PTFE with encapsulated steel spring to provide firm grip
  • Clips are numbered with joint size for easy identification
  • Can be used at temperatures up to 250°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 Pk.
PTFE
Conical Joints
250°C
Joint Clip
Encapsulated Steel spring
24/29
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Quickfit™ PTFE Joint Clips with Conical Joints > For use with: 24/29 conical joint glassware