Learn More
Glindemann™ PTFE Sealing Rings for taper joints (laboratory glassware)
Click to view available options
Size:
NS 10
NS 12
NS 14
NS 19
NS 24
NS 29
NS 34
NS 45
NS 7
Unit Size:
Pack of 50
Description
- Air leak rate <10-8 to 10-6 mbar l/s (high vacuum, overpressure).
- Solvent leak rate in containers <0.1 to 0.03mg/d.
- Temperature resistant from -200 to +300°C.
- The elastic ring (0.07mm thick) adheres to the ground joint without a groove, is reusable and fits to ground joint clamps.
- Other sizes on request.
Specifications
Specifications
| Material | PTFE |
| Size | NS 10 |