Learn More
PTGS1 293T Cell Overexpression Lysate (Denatured), Abnova
Product Code. 16189871
Click to view available options
:
100μL
Product Code. 16189871
Brand: Abnova H00005742T01.100uL
Please to purchase this item. Need a web account? Register with us today!
Western Blotting
Specifications
Cell Line 293T
For Use With (Application) Western Blot
Description PTGS1 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. BC029840.1
Molecular Weight (g/mol) 68.7
pH Range 6.8
Plasmid pCMV-PTGS1 full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-PTGS1 antibody (H00005742-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.