missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ PU 23

Product Code. p-7166812
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18776526

Brand: Tocris™ 6885/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Multidrug resistance protein (MRP) 4 inhibitor

PU 23 is a multidrug resistance protein (MRP) 4 inhibitor. Sensitizes MRP4-overexpressing cells to the anticancer agent 6-Mercaptopurine (Cat. No. 4103; 6-MP) (IC50 values are 14.11 and 6.06 μM in the absence and presence of compound, respectively). Increases accumulation of 6-MP in MRP4-overexpressing cells.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Molecular Formula C21H19N3O3S2
Formula Weight 425.52
Product Type Inhibitor
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.