Learn More
BUCHI Pure Glass Tubes
Suitable for a variety of applications
Brand:  BUCHI 148623411
 View more versions of this product
Product Code. 17203334
  • 571.00 € / Pack of 500
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Pure glass tubes
  • Suitable for a variety of applications
Specifications
Specifications
150 mm
Glass
Test Tubes
25 mm