Learn More
Masterflex™ PVC High Pressure Tubing with Luer Ends
Product Code. 12606225
Click to view available options
Overall Length (Metric):
1.21 mm
Unit Size:
Pack of 10
Product Code. 12606225
Brand: Masterflex™ 3052618
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Material PVC
Overall Length (Metric) 1.21 mm
Packaging Quantity 10 Pack
Sterility Non-sterile
Maximum Pressure 900 psi