Learn More
Cytiva PVC Tubing
Tubing for Peristaltic Pump P-1.
Brand: Cytiva 19-7629-01
This item is not returnable.
View return policy
Description
Specifications
Peristaltic Pump p-1 | |
1 mm | |
PVC |
Tubing | |
140 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva PVC Tubing > I.D.: 1mm; Length: 140mm
Spot an opportunity for improvement?Share a Content Correction