Learn More
Cole-Parmer™ PVDF Manifold, 2 port
Product Code. 11735278
Click to view available options
Tubing Size:
1/2 in. x 3/8 in.
1/4 in. x 1/8 in.
3/8 in. x 1/4 in.
Unit Size:
Each
Product Code. 11735278
Brand: Cole-Parmer™ 3152331
Please to purchase this item. Need a web account? Register with us today!
Expand your flow and line capacity with these manifolds. Choose the number of ports and connections to suit your needs.
  • Constructed of durable PVDF.
  • Ideal for applications requiring excellent chemical compatibly.
  • Available in a variety of configurations
Specifications
Material PVDF
Overall Length (English) 1.75 in.
Overall Length (Metric) 4.45 cm
Min. Operating Temperature (English) -65°F
Min. Operating Temperature (Metric) -54°C
Tubing Size 1/4 in. x 1/8 in.
Inlet 1.25 in. NPT(F)
Outlet Port 1/8 in. NPT(F)
Width (Metric) 2.54 cm
Width (English) 1 in.
Height (Metric) 2.54 cm
Height (English) 1 in.
Maximum Pressure 110 psi
Max. Operating Temperature (English) 230°F
Max. Operating Temperature (Metric) 110°C
Product Type Manifold