Learn More
Invitrogen™ PXK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554970
Description
Immunogen sequence: LSLPLPPKKL IGNMDREFIA ERQKGLQNYL NVITTNHILS NCELVKKFLD PNNYSANYTE IALQQVSMFF RSEPKWEVVE PLKDIGWRIR KKYFLMKIKN QPKERLVLSW ADLGPDKYLS DKDFQCLIKL LP Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
PXK is widely expressed in all tissues examined except in heart. It binds to and modulates brain Na,K-ATPase subunits ATP1B1 and ATP1B3 and may thereby participate in the regulation of electrical excitability and synaptic transmission. However, the protein kinase domain is predicted to be catalytically inactive.
Specifications
| PXK | |
| Polyclonal | |
| Unconjugated | |
| PXK | |
| C230080L11Rik; D14Ertd813e; EMBL:AAP92800.1, ECO:0000312; modulator of Na,K-ATPase; modulator of Na,K-ATPase long form; MONaKA; PX domain containing serine/threonine kinase; PX domain containing serine/threonine kinase like; PX domain-containing protein kinase-like protein; PX ser/thr kinase v2; PX serine/threonine kinase; pxk; pxk {ECO:0000312; RGD:727819} | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54899 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z7A4 | |
| PXK | |
| Recombinant protein corresponding to Human PXK. Recombinant protein control fragment (Product #RP-91475). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.